siRNA Details
Gene Symbol: SEZ6L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114099.2 | NP_001107571.1 | 4 | 568 | ||
NM_001243332.1 | NP_001230261.1 | 4 | 778 | |||
NM_001243333.1 | NP_001230262.1 | 3 | 646 | |||
NM_012410.3 | NP_036542.1 | 4 | 568 | |||
NM_201575.3 | NP_963869.2 | 4 | 778 | |||
XM_005255252.2 | XP_005255309.1 | 4 | 778 | |||
GenBank mRNA | AB105376.1 | - | 4 | 407 | ||
AJ245820.1 | - | 4 | 407 | |||
AJ245821.1 | - | 4 | 407 | |||
AJ245822.1 | - | 4 | 407 | |||
AK027407.1 | - | 2 | 48 | |||
AK027676.1 | - | 1 | 18 | |||
AK293675.1 | - | 3 | 491 | |||
AK299103.1 | - | 4 | 775 | |||
AY358404.1 | - | 4 | 767 | |||
KJ534938.1 | - | 4 | 533 | |||
KJ534940.1 | - | 4 | 533 | |||
KJ534988.1 | - | 4 | 533 | |||
KJ535068.1 | - | 4 | 533 | |||
KJ535092.1 | - | 4 | 532 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
