siRNA Details
Gene Symbol: OR8B8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012378.1 | NP_036510.1 | 1 | 195 | ||
GenBank mRNA | AF238488.1 | - | 1 | 195 | ||
BC130393.1 | - | 1 | 268 | 40146424 | ||
BC130419.1 | - | 1 | 268 | 40146450 |
