siRNA Details
Gene Symbol: OR12D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013936.3 | NP_039224.2 | 1 | 220 | ||
GenBank mRNA | BC069123.1 | - | 1 | 159 | 7216978 | |
BC101742.1 | - | 1 | 220 | 8069248 | ||
BC101746.1 | - | 1 | 220 | 8069252 |
