siRNA Details
Gene Symbol: HAVCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF043724.1 | - | 4 | 571 | ||
AK298259.1 | - | 4 | 852 | |||
HQ412639.1 | - | 3 | 520 | |||
HQ412640.1 | - | 3 | 520 | |||
HQ412641.1 | - | 3 | 520 | |||
JX049978.1 | - | 3 | 520 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
