siRNA Details
Gene Symbol: TCL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028288.1 | 5 | 1490 | |||
GenBank mRNA | AB035332.1 | - | 2 | 3269 | ||
AB035333.1 | - | 2 | 3269 | |||
AB035334.1 | - | 3 | 2301 | |||
AB035335.1 | - | 2 | 1846 | |||
AB035337.1 | - | 5 | 1492 | |||
AB035338.1 | - | 5 | 1687 | |||
AF195821.1 | - | 1 | 92 | |||
BC041075.1 | - | 2 | 1660 | 5923478 | ||
BX161462.1 | - | 7 | 1535 |
