siRNA Details
Gene Symbol: MLH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040108.1 | NP_001035197.1 | 2 | 2858 | ||
NM_014381.2 | NP_055196.2 | 2 | 2858 | |||
XM_005267532.4 | XP_005267589.1 | 2 | 2858 | |||
XM_005267533.4 | XP_005267590.1 | 2 | 2858 | |||
XM_005267534.2 | XP_005267591.1 | 2 | 2858 | |||
XM_006720116.3 | XP_006720179.1 | 2 | 2770 | |||
XM_011536646.2 | XP_011534948.1 | 2 | 2858 | |||
XM_011536648.2 | XP_011534950.1 | 2 | 2858 | |||
XM_017021219.1 | XP_016876708.1 | 2 | 2784 | |||
GenBank mRNA | AB039667.1 | - | 1 | 2642 | ||
AF195657.1 | - | 2 | 2756 | |||
AL833875.1 | - | 1 | 688 | |||
BC112167.1 | - | 1 | 2678 | 8327635 | ||
DA731365.1 | - | 1 | 675 |
