siRNA Details
Gene Symbol: PDLIM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257962.1 | NP_001244891.1 | 3, 4 | 462 | ||
NM_014476.5 | NP_055291.2 | 3, 4 | 462 | |||
GenBank mRNA | AF002280.1 | - | 3, 4 | 384 | ||
AF039018.1 | - | 3, 4 | 408 | |||
AK222966.1 | - | 3, 4 | 366 | |||
AK313253.1 | - | 3, 4 | 411 | |||
BX647263.1 | - | 1 | 3390 | |||
C03422.1 | - | 3, 4 | 288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
