siRNA Details
Gene Symbol: PDLIM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014476.5 | NP_055291.2 | 5 | 694 | ||
GenBank mRNA | AF002280.1 | - | 5 | 616 | ||
AF039018.1 | - | 5 | 640 | |||
AK222966.1 | - | 5 | 598 | |||
AK313253.1 | - | 5 | 643 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
