siRNA Details
Gene Symbol: ZNF544 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320767.1 | NP_001307696.1 | 7 | 1715 | ||
NM_001320769.1 | NP_001307698.1 | 7 | 1757 | |||
NM_001320770.1 | NP_001307699.1 | 6 | 1631 | |||
NM_001320771.1 | NP_001307700.1 | 6 | 1596 | |||
NM_001320773.1 | NP_001307702.1 | 6 | 1673 | |||
NM_001320774.1 | NP_001307703.1 | 7 | 1052 | |||
NM_001320777.1 | NP_001307706.1 | 8 | 1286 | |||
NM_001320781.1 | NP_001307710.1 | 8 | 2068 | |||
NM_001320785.1 | NP_001307714.1 | 7 | 940 | |||
NM_001320786.1 | NP_001307715.1 | 7 | 975 | |||
NM_001320787.1 | NP_001307716.1 | 7 | 1017 | |||
NM_001320788.1 | NP_001307717.1 | 7 | 1645 | |||
NM_001320789.1 | NP_001307718.1 | 7 | 1722 | |||
NM_001320791.1 | NP_001307720.1 | 7 | 1680 | |||
NM_001320792.1 | NP_001307721.1 | 8 | 1731 | |||
NM_014480.3 | NP_055295.2 | 7 | 1680 | |||
XM_011526738.2 | XP_011525040.1 | 8 | 1793 | |||
XM_017026603.1 | XP_016882092.1 | 8 | 1758 | |||
GenBank mRNA | AF020591.1 | - | 6 | 1428 | ||
AK226080.1 | - | 8 | 2439 | |||
AK290297.1 | - | 5 | 1126 | |||
AK291656.1 | - | 7 | 1432 | |||
AK296846.1 | - | 5 | 1341 | |||
AK301650.1 | - | 8 | 692 | |||
AK316446.1 | - | 6 | 1383 | |||
AL833618.1 | - | 7 | 1409 | |||
BC067271.1 | - | 6 | 1408 | 6199648 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
