siRNA Details
Gene Symbol: ZNF544 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320767.1 | NP_001307696.1 | 7 | 2135 | ||
NM_001320769.1 | NP_001307698.1 | 7 | 2177 | |||
NM_001320770.1 | NP_001307699.1 | 6 | 2051 | |||
NM_001320771.1 | NP_001307700.1 | 6 | 2016 | |||
NM_001320773.1 | NP_001307702.1 | 6 | 2093 | |||
NM_001320774.1 | NP_001307703.1 | 7 | 1472 | |||
NM_001320777.1 | NP_001307706.1 | 8 | 1706 | |||
NM_001320781.1 | NP_001307710.1 | 8 | 2488 | |||
NM_001320785.1 | NP_001307714.1 | 7 | 1360 | |||
NM_001320786.1 | NP_001307715.1 | 7 | 1395 | |||
NM_001320787.1 | NP_001307716.1 | 7 | 1437 | |||
NM_001320788.1 | NP_001307717.1 | 7 | 2065 | |||
NM_001320789.1 | NP_001307718.1 | 7 | 2142 | |||
NM_001320791.1 | NP_001307720.1 | 7 | 2100 | |||
NM_001320792.1 | NP_001307721.1 | 8 | 2151 | |||
NM_014480.3 | NP_055295.2 | 7 | 2100 | |||
XM_011526738.2 | XP_011525040.1 | 8 | 2213 | |||
XM_017026603.1 | XP_016882092.1 | 8 | 2178 | |||
GenBank mRNA | AF020591.1 | - | 6 | 1848 | ||
AK226080.1 | - | 8 | 2859 | |||
AK290297.1 | - | 5 | 1546 | |||
AK291656.1 | - | 7 | 1852 | |||
AK296846.1 | - | 5 | 1761 | |||
AK316446.1 | - | 6 | 1803 | |||
AL833618.1 | - | 7 | 1829 | |||
BC067271.1 | - | 6 | 1828 | 6199648 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
