siRNA Details
Gene Symbol: PPP2R3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013239.4 | NP_037371.2 | 11 | 1703 | ||
XM_011545630.2 | XP_011543932.1 | 11 | 1703 | |||
GenBank mRNA | AB209137.1 | - | 7 | 1309 | ||
AF135016.1 | - | 10 | 1067 | |||
AF155098.1 | - | 5 | 478 | |||
AL157501.1 | - | 3 | 414 | |||
BC009032.1 | - | 3 | 660 | |||
BC011180.1 | - | 5 | 480 | |||
BC063429.1 | - | 11 | 1458 | 5239974 | ||
BF530463.1 | - | 3 | 667 | |||
BM715684.1 | - | 5 | 443 |
