siRNA Details
Gene Symbol: FAM156B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099684.2 | NP_001093154.1 | 3 | 1102 | ||
NM_001321178.1 | NP_001308107.1 | 3 | 2868 | |||
NM_001321179.1 | NP_001308108.1 | 4 | 2127 | |||
NM_001321180.1 | NP_001308109.1 | 4 | 1265 | |||
NM_001321181.1 | NP_001308110.1 | 4 | 1237 | |||
NM_001321182.1 | NP_001308111.1 | 3 | 1126 | |||
NM_001321183.1 | NP_001308112.1 | 4 | 946 | |||
NM_001321186.1 | NP_001308115.1 | 4 | 828 | |||
NM_001321187.1 | NP_001308116.1 | 4 | 810 | |||
NM_001321188.1 | NP_001308117.1 | 4 | 754 | |||
XM_011530817.1 | XP_011529119.1 | 4 | 1219 | |||
XM_011530821.2 | XP_011529123.1 | 3 | 727 | |||
GenBank mRNA | AK091598.1 | - | 4 | 2023 | ||
BX649013.1 | - | 3 | 2831 |
Gene Symbol: FAM156A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242489.1 | NP_001229418.1 | 3 | 1179 | ||
NM_001242490.1 | NP_001229419.1 | 4 | 1163 | |||
NM_001242491.1 | NP_001229420.1 | 3 | 1126 | |||
NM_001242492.1 | NP_001229421.1 | 5 | 1012 | |||
NM_001242493.1 | NP_001229422.1 | 5 | 905 | |||
NM_001242494.1 | NP_001229423.1 | 4 | 863 | |||
NM_001242495.1 | NP_001229424.1 | 3 | 770 | |||
NM_001242496.1 | NP_001229425.1 | 4 | 754 | |||
NM_001242497.1 | NP_001229426.1 | 3 | 661 | |||
NM_014138.4 | NP_054857.2 | 5 | 1314 | |||
XM_006724586.3 | XP_006724649.1 | 4 | 2411 | |||
XM_006724587.3 | XP_006724650.1 | 4 | 2002 | |||
XM_011530781.1 | XP_011529083.1 | 4 | 1219 | |||
XM_011530782.1 | XP_011529084.1 | 4 | 1488 | |||
XM_011530783.1 | XP_011529085.1 | 4 | 946 | |||
XM_011530784.2 | XP_011529086.1 | 3 | 715 | |||
XM_017029457.1 | XP_016884946.1 | 5 | 3170 | |||
XM_017029458.1 | XP_016884947.1 | 5 | 2761 | |||
GenBank mRNA | AF090943.1 | - | 1 | 354 | ||
AF370413.1 | - | 3 | 1328 | |||
AK054721.1 | - | 3 | 1740 | |||
AK057692.1 | - | 3 | 1109 | |||
AK092365.1 | - | 4 | 1458 | |||
AK096162.1 | - | 3 | 1614 | |||
AL834135.1 | - | 3 | 778 | |||
BC000867.1 | - | 1 | 329 | 3460178 | ||
BC067819.1 | - | 3 | 762 | 5313567 | ||
CR749619.1 | - | 2 | 619 |
