siRNA Details
Gene Symbol: ENTPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001248.3 | NP_001239.2 | 7, 8 | 954 | ||
NM_001291960.1 | NP_001278889.1 | 7, 8 | 995 | |||
NM_001291961.1 | NP_001278890.1 | 7, 8 | 954 | |||
GenBank mRNA | AF034840.2 | - | 7, 8 | 915 | ||
AF039917.1 | - | 7, 8 | 918 | |||
AK313322.1 | - | 7, 8 | 980 | |||
BC029869.1 | - | 6, 7 | 858 | 5174918 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
