siRNA Details
Gene Symbol: POMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013382.5 | NP_037514.2 | 13 | 1627 | ||
XM_011536675.2 | XP_011534977.1 | 13 | 1627 | |||
XM_011536676.2 | XP_011534978.1 | 12 | 1540 | |||
XM_011536677.2 | XP_011534979.1 | 9 | 1169 | |||
XM_011536679.2 | XP_011534981.1 | 9 | 829 | |||
XM_017021244.1 | XP_016876733.1 | 12 | 1461 | |||
XM_017021245.1 | XP_016876734.1 | 9 | 915 | |||
GenBank mRNA | AB209114.1 | - | 13 | 1449 | ||
AF105020.1 | - | 13 | 1630 | |||
AK023804.1 | - | 7 | 2218 | |||
AK025962.1 | - | 3 | 246 | |||
AY090480.1 | - | 13 | 1630 | |||
BC031651.1 | - | 13 | 1517 | 5169145 | ||
BE797202.1 | - | 33 | ||||
CR627447.1 | - | 13 | 1197 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
