siRNA Details
Gene Symbol: CD209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144893.1 | NP_001138365.1 | 5 | 758 | ||
NM_001144894.1 | NP_001138366.1 | 5 | 1034 | |||
NM_001144895.1 | NP_001138367.1 | 7 | 890 | |||
NM_001144896.1 | NP_001138368.1 | 6 | 1094 | |||
NM_001144897.1 | NP_001138369.1 | 7 | 1148 | |||
NM_001144899.1 | NP_001138371.1 | 7 | 683 | |||
NM_021155.3 | NP_066978.1 | 7 | 1166 | |||
NR_026692.1 | 6 | 1266 | ||||
XM_005272472.4 | XP_005272529.1 | 7 | 783 | |||
GenBank mRNA | AF290886.1 | - | 7 | 1108 | ||
AK293089.1 | - | 7 | 1122 | |||
AK304190.1 | - | 7 | 669 | |||
AK308595.1 | - | 4 | 709 | |||
AK313585.1 | - | 7 | 1167 | |||
AY042221.1 | - | 7 | 1099 | |||
AY042222.1 | - | 7 | 1081 | |||
AY042223.1 | - | 7 | 823 | |||
AY042224.1 | - | 6 | 391 | |||
AY042225.1 | - | 6 | 1027 | |||
AY042226.1 | - | 5 | 967 | |||
AY042227.1 | - | 6 | 691 | |||
AY042229.1 | - | 6 | 1199 | |||
AY042230.1 | - | 5 | 1127 | |||
AY042231.1 | - | 5 | 989 | |||
BC110615.2 | - | 5 | 1134 | 40032459 | ||
M98457.1 | - | 7 | 1140 |
