siRNA Details
Gene Symbol: CYHR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129888.1 | NP_001123360.1 | 3 | 1101 | ||
NM_032687.3 | NP_116076.1 | 3 | 1137 | |||
XM_006716560.3 | XP_006716623.1 | 3 | 1354 | |||
XM_006716561.3 | XP_006716624.1 | 3 | 1306 | |||
GenBank mRNA | BC005073.1 | - | 3 | 1137 | 3533499 | |
BC056412.1 | - | 2 | 1415 | |||
BC065484.1 | - | 3 | 1101 | 4638402 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
