siRNA Details
Gene Symbol: COPS7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164093.1 | NP_001157565.1 | 3, 4 | 702 | ||
NM_001164094.1 | NP_001157566.1 | 4, 5 | 567 | |||
NM_001164095.2 | NP_001157567.1 | 4, 5 | 638 | |||
NM_016319.3 | NP_057403.1 | 4, 5 | 540 | |||
XM_005253694.2 | XP_005253751.1 | 4, 5 | 465 | |||
GenBank mRNA | AB014764.1 | - | 3, 4 | 328 | ||
AB033603.1 | - | 4, 5 | 405 | |||
AF193844.1 | - | 3, 4 | 328 | |||
AF210052.1 | - | 4, 5 | 469 | |||
AK001318.1 | - | 4, 5 | 452 | |||
AK021721.1 | - | 4, 5 | 505 | |||
AK022488.1 | - | 4, 5 | 447 | |||
AK292621.1 | - | 3, 4 | 698 | |||
BC011789.2 | - | 4, 5 | 428 | 4025790 | ||
BC093015.1 | - | 4, 5 | 479 | 30344094 | ||
DA365275.1 | - | 4, 5 | 512 |
