siRNA Details
Gene Symbol: CCDC53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301107.1 | NP_001288036.1 | 7 | 701 | ||
NM_016053.3 | NP_057137.1 | 7 | 704 | |||
XM_017019383.1 | XP_016874872.1 | 6 | 519 | |||
GenBank mRNA | AF151874.1 | - | 7 | 649 | ||
AF155655.1 | - | 7 | 649 | |||
AK223339.1 | - | 7 | 640 | |||
AK314970.1 | - | 7 | 622 | |||
BC010889.1 | - | 8 | 575 | 4272385 | ||
BX356987.2 | - | 7 | 656 | |||
CR457171.1 | - | 7 | 542 |
