siRNA Details
Gene Symbol: TFB1M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016020.3 | NP_057104.2 | 1 | 113 | ||
XM_005267006.4 | XP_005267063.1 | 1 | 112 | |||
XM_011535870.1 | XP_011534172.1 | 1 | 127 | |||
XM_011535871.1 | XP_011534173.1 | 1 | 127 | |||
XM_011535872.2 | XP_011534174.1 | 1 | 127 | |||
GenBank mRNA | AF151833.1 | - | 1 | 130 | ||
AK289484.1 | - | 1 | 127 | |||
BC005183.1 | - | 1 | 111 | |||
BC017788.1 | - | 1 | 113 | 4672440 | ||
BC054007.1 | - | 1 | 118 | |||
BU849912.1 | - | 1 | 122 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
