siRNA Details
Gene Symbol: TRIM17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024940.2 | NP_001020111.1 | 7 | 1940 | ||
NM_016102.3 | NP_057186.1 | 7 | 1976 | |||
XM_006711779.2 | XP_006711842.1 | 7 | 2003 | |||
XM_011544209.2 | XP_011542511.1 | 7 | 1990 | |||
XM_011544210.2 | XP_011542512.1 | 7 | 2092 | |||
XM_011544211.2 | XP_011542513.1 | 7 | 1993 | |||
XM_017001418.1 | XP_016856907.1 | 5 | 2964 | |||
XM_017001419.1 | XP_016856908.1 | 6 | 2508 | |||
GenBank mRNA | AK055426.1 | - | 6 | 2544 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
