siRNA Details
Gene Symbol: ING4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127582.1 | NP_001121054.1 | 2 | 136 | ||
NM_001127583.1 | NP_001121055.1 | 2 | 136 | |||
NM_001127584.1 | NP_001121056.1 | 2 | 136 | |||
NM_001127586.1 | NP_001121058.1 | 2 | 136 | |||
NM_016162.3 | NP_057246.2 | 2 | 136 | |||
XM_005253698.3 | XP_005253755.1 | 2 | 108 | |||
XM_011520965.2 | XP_011519267.1 | 2 | 83 | |||
XM_017019392.1 | XP_016874881.1 | 2 | 108 | |||
XM_017019393.1 | XP_016874882.1 | 2 | 108 | |||
GenBank mRNA | AB197695.1 | - | 2 | 95 | ||
AB197696.1 | - | 2 | 95 | |||
AB197697.1 | - | 2 | 95 | |||
AF063594.1 | - | 2 | 114 | |||
AF110645.1 | - | 2 | 112 | |||
AF156552.1 | - | 2 | 112 | |||
AK303354.1 | - | 2 | 118 | |||
AK313621.1 | - | 2 | 108 | |||
BC007781.2 | - | 2 | 98 | 4309278 | ||
BC013038.2 | - | 2 | 81 | |||
BC095434.1 | - | 2 | 100 | 30365729 | ||
BG705367.1 | - | 2 | 136 | |||
BG718170.1 | - | 2 | 147 | |||
BM922497.1 | - | 2 | 89 | |||
BQ066994.1 | - | 107 | ||||
EF152350.1 | - | 2 | 95 | |||
EF152351.1 | - | 2 | 95 | |||
EF152352.1 | - | 2 | 95 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
