siRNA Details
Gene Symbol: ING4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127582.1 | NP_001121054.1 | 6 | 632 | ||
NM_001127583.1 | NP_001121055.1 | 6 | 623 | |||
NM_001127584.1 | NP_001121056.1 | 6 | 620 | |||
NM_001127585.1 | NP_001121057.1 | 5 | 560 | |||
NM_016162.3 | NP_057246.2 | 6 | 629 | |||
XM_005253698.3 | XP_005253755.1 | 7 | 880 | |||
XM_011520964.2 | XP_011519266.1 | 5 | 558 | |||
XM_011520965.2 | XP_011519267.1 | 6 | 579 | |||
XM_017019392.1 | XP_016874881.1 | 7 | 877 | |||
XM_017019393.1 | XP_016874882.1 | 7 | 868 | |||
GenBank mRNA | AB197695.1 | - | 6 | 588 | ||
AB197696.1 | - | 6 | 582 | |||
AB197697.1 | - | 6 | 579 | |||
AF063594.1 | - | 6 | 607 | |||
AF110645.1 | - | 6 | 608 | |||
AF156552.1 | - | 6 | 608 | |||
AK293169.1 | - | 5 | 545 | |||
AK313621.1 | - | 6 | 601 | |||
BC007781.2 | - | 6 | 594 | 4309278 | ||
BC013038.2 | - | 6 | 574 | |||
BC095434.1 | - | 6 | 593 | 30365729 | ||
BG718170.1 | - | 6 | 640 | |||
BM922497.1 | - | 7 | 858 | |||
EF152349.1 | - | 5 | 519 | |||
EF152350.1 | - | 5 | 424 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
