siRNA Details
Gene Symbol: C3orf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171740.2 | NP_001165211.1 | 5 | 1025 | ||
NM_001171741.2 | NP_001165212.1 | 5 | 820 | |||
NM_001171743.2 | NP_001165214.2 | 5 | 804 | |||
NM_016210.4 | NP_057294.2 | 6 | 1114 | |||
XM_011533781.2 | XP_011532083.1 | 4 | 1764 | |||
XM_011533782.2 | XP_011532084.1 | 6 | 1102 | |||
XM_011533783.2 | XP_011532085.1 | 5 | 992 | |||
XM_011533784.2 | XP_011532086.1 | 6 | 880 | |||
XM_011533785.2 | XP_011532087.1 | 5 | 818 | |||
XM_011533788.1 | XP_011532090.1 | 4 | 424 | |||
XM_011533789.1 | XP_011532091.1 | 3 | 449 | |||
XM_011533790.1 | XP_011532092.1 | 6 | 1030 | |||
XM_017006545.1 | XP_016862034.1 | 5 | 942 | |||
XM_017006546.1 | XP_016862035.1 | 4 | 269 | |||
XM_017006547.1 | XP_016862036.1 | 5 | 965 | |||
GenBank mRNA | AF188706.1 | - | 5 | 785 | ||
AK124438.1 | - | 5 | 865 | |||
AK308074.1 | - | 5 | 686 | |||
AK309486.1 | - | 4 | 584 | |||
AK311093.1 | - | 2 | 305 | |||
AL833938.1 | - | 6 | 1073 | |||
BC034766.1 | - | 5 | 898 | 5177784 | ||
DA535127.1 | - | 4 | 460 | |||
DB027651.1 | - | 4 | 424 |
