siRNA Details
Gene Symbol: C3orf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171740.2 | NP_001165211.1 | 5 | 1800 | ||
NM_001171741.2 | NP_001165212.1 | 5 | 1595 | |||
NM_001171743.2 | NP_001165214.2 | 5 | 1579 | |||
NM_016210.4 | NP_057294.2 | 6 | 1889 | |||
XM_011533781.2 | XP_011532083.1 | 4 | 2539 | |||
XM_011533782.2 | XP_011532084.1 | 6 | 1877 | |||
XM_011533783.2 | XP_011532085.1 | 5 | 1767 | |||
XM_011533784.2 | XP_011532086.1 | 6 | 1655 | |||
XM_011533785.2 | XP_011532087.1 | 5 | 1593 | |||
XM_011533788.1 | XP_011532090.1 | 4 | 1199 | |||
XM_011533789.1 | XP_011532091.1 | 3 | 1224 | |||
XM_011533790.1 | XP_011532092.1 | 6 | 1805 | |||
XM_017006545.1 | XP_016862034.1 | 5 | 1717 | |||
XM_017006546.1 | XP_016862035.1 | 4 | 1044 | |||
XM_017006547.1 | XP_016862036.1 | 5 | 1740 | |||
GenBank mRNA | AF188706.1 | - | 5 | 1560 | ||
AK124438.1 | - | 5 | 1640 | |||
AK130227.1 | - | 3 | 995 | |||
AK308074.1 | - | 5 | 1461 | |||
AL833938.1 | - | 6 | 1848 | |||
BC034766.1 | - | 5 | 1673 | 5177784 |
