siRNA Details
Gene Symbol: DCTN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135643.1 | NP_001129115.1 | 1 | 197 | ||
NM_016221.3 | NP_057305.1 | 1 | 197 | |||
XM_005268457.1 | XP_005268514.1 | 1 | 135 | |||
XM_011537646.1 | XP_011535948.1 | 1 | 197 | |||
GenBank mRNA | AF195120.1 | - | 1 | 112 | ||
AK000299.1 | - | 1 | 116 | |||
AK296787.1 | - | 1 | 116 | |||
AK307757.1 | - | 1 | 136 | |||
AK311693.1 | - | 1 | 133 | |||
BC026323.1 | - | 1 | 122 | 4815273 | ||
DC321382.1 | - | 1 | 197 | |||
KU178637.1 | - | 1 | 95 | |||
KU178638.1 | - | 1 | 95 | |||
KU178639.1 | - | 1 | 95 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
