siRNA Details
Gene Symbol: DCTN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135643.1 | NP_001129115.1 | 12 | 1112 | ||
NM_001135644.1 | NP_001129116.1 | 11 | 1259 | |||
NM_016221.3 | NP_057305.1 | 11 | 1091 | |||
XM_005268457.1 | XP_005268514.1 | 10 | 985 | |||
XM_011537644.1 | XP_011535946.1 | 12 | 995 | |||
XM_011537645.1 | XP_011535947.1 | 12 | 907 | |||
XM_011537646.1 | XP_011535948.1 | 12 | 1128 | |||
XM_011537647.1 | XP_011535949.1 | 12 | 951 | |||
GenBank mRNA | AF195120.1 | - | 11 | 1006 | ||
AK000299.1 | - | 11 | 1010 | |||
AK125973.1 | - | 11 | 1259 | |||
AK299517.1 | - | 11 | 1012 | |||
AK307757.1 | - | 12 | 1051 | |||
AL162085.1 | - | 1 | 41 | |||
BC026323.1 | - | 11 | 1016 | 4815273 | ||
KU178637.1 | - | 11 | 989 | |||
KU178638.1 | - | 11 | 989 | |||
KU178639.1 | - | 7 | 745 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
