siRNA Details
Gene Symbol: NUSAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243142.1 | NP_001230071.1 | 9 | 1282 | ||
NM_001243143.1 | NP_001230072.1 | 9 | 1243 | |||
NM_001301136.1 | NP_001288065.1 | 9 | 1285 | |||
NM_016359.4 | NP_057443.2 | 9 | 1288 | |||
NM_018454.7 | NP_060924.4 | 9 | 1285 | |||
XM_005254428.2 | XP_005254485.1 | 9 | 1157 | |||
XM_005254430.4 | XP_005254487.1 | 9 | 1079 | |||
XM_006720559.2 | XP_006720622.1 | 9 | 1160 | |||
XM_006720560.3 | XP_006720623.1 | 9 | 1256 | |||
XM_006720561.2 | XP_006720624.1 | 9 | 1074 | |||
XM_017022295.1 | XP_016877784.1 | 9 | 1952 | |||
GenBank mRNA | AF090915.1 | - | 9 | 1091 | ||
AF290612.1 | - | 9 | 1092 | |||
AF305711.1 | - | 9 | 1021 | |||
AK023483.1 | - | 9 | 1095 | |||
AK222819.1 | - | 9 | 1109 | |||
AK225703.1 | - | 9 | 1099 | |||
AK290879.1 | - | 9 | 1112 | |||
AK307728.1 | - | 9 | 1082 | |||
AL833611.1 | - | 9 | 1055 | |||
BC001308.1 | - | 9 | 1088 | 3453354 | ||
BC010838.1 | - | 7 | 829 | |||
BC012887.1 | - | 9 | 1066 | 3882061 | ||
BC024772.1 | - | 9 | 1090 | 4342499 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
