siRNA Details
Gene Symbol: KLF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016531.5 | NP_057615.3 | 5 | 1065 | ||
XM_017008279.1 | XP_016863768.1 | 6 | 1259 | |||
GenBank mRNA | AB024523.1 | - | 5 | 788 | ||
AF285837.1 | - | 5 | 892 | |||
AK313459.1 | - | 5 | 1035 | |||
BC030662.1 | - | 5 | 1040 | 5262512 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
