siRNA Details
Gene Symbol: ZDHHC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016598.2 | NP_057682.1 | 5, 6 | 893 | ||
XM_005265209.1 | XP_005265266.1 | 5, 6 | 893 | |||
XM_006713192.2 | XP_006713255.1 | 6, 7 | 995 | |||
XM_011533797.1 | XP_011532099.1 | 6, 7 | 995 | |||
XM_011533800.2 | XP_011532102.1 | 6, 7 | 988 | |||
XM_017006557.1 | XP_016862046.1 | 6, 7 | 995 | |||
XM_017006558.1 | XP_016862047.1 | 7, 8 | 1041 | |||
XM_017006559.1 | XP_016862048.1 | 6, 7 | 1090 | |||
XM_017006561.1 | XP_016862050.1 | 5, 6 | 893 | |||
XM_017006563.1 | XP_016862052.1 | 6, 7 | 995 | |||
XM_017006564.1 | XP_016862053.1 | 6, 7 | 995 | |||
XM_017006565.1 | XP_016862054.1 | 6, 7 | 976 | |||
XM_017006568.1 | XP_016862057.1 | 4, 5 | 402 | |||
XM_017006571.1 | XP_016862060.1 | 5, 6 | 889 | |||
GenBank mRNA | AF247703.1 | - | 5, 6 | 833 | ||
AY605661.1 | - | 3, 4 | 313 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
