siRNA Details
Gene Symbol: FAM13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101800.2 | NP_001095270.1 | 3 | 577 | ||
NM_016603.3 | NP_057687.2 | 3 | 577 | |||
XM_005272007.1 | XP_005272064.1 | 3 | 577 | |||
XM_006714647.1 | XP_006714710.1 | 3 | 577 | |||
XM_006714648.1 | XP_006714711.1 | 3 | 329 | |||
XM_011543448.1 | XP_011541750.1 | 4 | 370 | |||
XM_011543449.1 | XP_011541751.1 | 4 | 674 | |||
XM_011543450.1 | XP_011541752.1 | 3 | 286 | |||
XM_017009549.1 | XP_016865038.1 | 3 | 602 | |||
XM_017009550.1 | XP_016865039.1 | 3 | 286 | |||
GenBank mRNA | AF251038.1 | - | 3 | 496 | ||
AK314224.1 | - | 3 | 553 | |||
BC048965.1 | - | 3 | 549 | 5265611 | ||
BC056887.1 | - | 2 | 152 | 6148274 | ||
BU183000.1 | - | 2 | 151 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
