siRNA Details
Gene Symbol: SPG21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127889.4 | NP_001121361.1 | 4 | 419 | ||
NM_016630.6 | NP_057714.1 | 4 | 600 | |||
XM_005254437.4 | XP_005254494.1 | 5 | 633 | |||
XM_017022297.1 | XP_016877786.1 | 4 | 968 | |||
XM_017022298.1 | XP_016877787.1 | 5 | 1058 | |||
GenBank mRNA | AF208861.1 | - | 3 | 315 | ||
AF212231.1 | - | 4 | 387 | |||
AK123725.1 | - | 2 | 832 | |||
AK172849.1 | - | 4 | 432 | |||
AK225527.1 | - | 4 | 432 | |||
AK312943.1 | - | 4 | 434 | |||
BC000244.1 | - | 4 | 470 | 3357388 | ||
BI552731.1 | - | 4 | 570 | |||
BI554234.1 | - | 5 | 630 | |||
BX446863.2 | - | 4 | 550 | |||
DA680695.1 | - | 4 | 418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
