siRNA Details
Gene Symbol: ZNF589 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016089.2 | NP_057173.2 | 4 | 2439 | ||
XM_017006575.1 | XP_016862064.1 | 4 | 2520 | |||
XM_017006576.1 | XP_016862065.1 | 6 | 2669 | |||
XM_017006577.1 | XP_016862066.1 | 4 | 3725 | |||
XM_017006578.1 | XP_016862067.1 | 4 | 2500 | |||
XM_017006579.1 | XP_016862068.1 | 6 | 2625 | |||
GenBank mRNA | AB209387.1 | - | 4 | 2420 | ||
AF114816.1 | - | 7 | 1458 | |||
AF114817.1 | - | 6 | 2742 | |||
AL832831.1 | - | 4 | 2426 | |||
AY258146.1 | - | 4 | 2439 | |||
BC005859.2 | - | 4 | 2398 | |||
BC006247.2 | - | 4 | 2419 | |||
BC008213.1 | - | 1 | 1007 | |||
BC048798.1 | - | 4 | 2416 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
