siRNA Details
Gene Symbol: ANAPC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137664.1 | NP_001131136.1 | 6 | 811 | ||
NM_016238.2 | NP_057322.2 | 6 | 811 | |||
XM_005253903.3 | XP_005253960.1 | 5 | 677 | |||
XM_011538454.2 | XP_011536756.1 | 6 | 1039 | |||
XM_017019425.1 | XP_016874914.1 | 6 | 811 | |||
XM_017019426.1 | XP_016874915.1 | 6 | 806 | |||
XM_017019427.1 | XP_016874916.1 | 6 | 1605 | |||
XM_017019428.1 | XP_016874917.1 | 5 | 694 | |||
GenBank mRNA | AF191340.1 | - | 6 | 708 | ||
AK021809.1 | - | 6 | 790 | |||
BC009498.2 | - | 4 | 381 | |||
BC017316.2 | - | 2 | 157 | |||
BC040130.1 | - | 6 | 726 | |||
BC098264.1 | - | 6 | 707 | |||
BC098295.1 | - | 6 | 707 | |||
BC098362.1 | - | 6 | 707 | |||
BC099732.1 | - | 6 | 707 | |||
BC141848.1 | - | 6 | 761 | 8860346 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
