siRNA Details
Gene Symbol: DCDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195610.1 | NP_001182539.1 | 4, 5 | 636 | ||
NM_016356.4 | NP_057440.2 | 3, 4 | 729 | |||
GenBank mRNA | AB032980.2 | - | 3, 4 | 427 | ||
AF181721.1 | - | 3, 4 | 729 | |||
AK027036.1 | - | 3, 4 | 545 | |||
BC050704.1 | - | 4, 5 | 636 | 5758845 | ||
BM923267.1 | - | 4, 5 | 649 |
