siRNA Details
Gene Symbol: ESF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276380.1 | NP_001263309.1 | 14 | 2512 | ||
NM_016649.3 | NP_057733.2 | 14 | 2523 | |||
XM_017027874.1 | XP_016883363.1 | 14 | 3975 | |||
GenBank mRNA | AK022670.1 | - | 2 | 313 | ||
AK074299.1 | - | 13 | 2009 |
