siRNA Details
Gene Symbol: HMP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015980.4 | NP_057064.1 | 3 | 410 | ||
GenBank mRNA | AF113537.1 | - | 3 | 279 | ||
AF123767.1 | - | 3 | 279 | |||
AF133422.1 | - | 3 | 263 | |||
AK295206.1 | - | 2 | 93 | |||
AK297196.1 | - | 3 | 294 | |||
AK312359.1 | - | 3 | 297 | |||
BC002619.2 | - | 3 | 293 | 3160345 | ||
BC018873.2 | - | 3 | 286 | |||
CR457142.1 | - | 2 | 148 | |||
DC366641.1 | - | 3 | 410 |
