siRNA Details
Gene Symbol: HMP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015980.4 | NP_057064.1 | 4, 5 | 580 | ||
GenBank mRNA | AF113537.1 | - | 4, 5 | 449 | ||
AF123767.1 | - | 4, 5 | 449 | |||
AF133422.1 | - | 4, 5 | 433 | |||
AK294383.1 | - | 3, 4 | 363 | |||
AK295206.1 | - | 3, 4 | 263 | |||
AK297196.1 | - | 4, 5 | 386 | |||
AK312359.1 | - | 4, 5 | 467 | |||
AK315949.1 | - | 2, 3 | 195 | |||
AK315952.1 | - | 3, 4 | 277 | |||
BC002619.2 | - | 4, 5 | 463 | 3160345 | ||
BC018873.2 | - | 4, 5 | 456 | |||
CR457142.1 | - | 3, 4 | 318 |
