siRNA Details
Gene Symbol: SARAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284239.1 | NP_001271168.1 | 2 | 590 | ||
NM_016127.5 | NP_057211.4 | 3 | 769 | |||
XM_017013567.1 | XP_016869056.1 | 3 | 1019 | |||
XM_017013568.1 | XP_016869057.1 | 3 | 447 | |||
XM_017013569.1 | XP_016869058.1 | 3 | 447 | |||
XM_017013570.1 | XP_016869059.1 | 2 | 529 | |||
GenBank mRNA | AB028926.1 | - | 3 | 629 | ||
AF078855.1 | - | 3 | 597 | |||
AF100748.1 | - | 3 | 526 | |||
AF490252.1 | - | 3 | 510 | |||
AK025927.1 | - | 3 | 679 | |||
AK075340.1 | - | 3 | 627 | |||
AK094386.1 | - | 4 | 610 | |||
AK130057.1 | - | 2 | 377 | |||
AK222632.1 | - | 3 | 526 | |||
AK295360.1 | - | 4 | 574 | |||
AK314659.1 | - | 3 | 651 | |||
AK315956.1 | - | 2 | 347 | |||
AY359104.1 | - | 3 | 694 | |||
AY454545.1 | - | 2 | 346 | |||
BC015012.1 | - | 3 | 598 | 3908758 | ||
JQ348891.1 | - | 3 | 638 | |||
JQ348892.1 | - | 2 | 330 |
