siRNA Details
Gene Symbol: CDKN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005192.3 | NP_005183.2 | 2 | 194 | ||
XM_011536353.2 | XP_011534655.1 | 2 | 210 | |||
GenBank mRNA | AF213033.1 | - | 1 | 80 | ||
AF213034.1 | - | 1 | 80 | |||
AF213035.1 | - | 1 | 80 | |||
AF213036.1 | - | 1 | 80 | |||
AF213037.1 | - | 1 | 80 | |||
AF213039.1 | - | 1 | 80 | |||
AF213040.1 | - | 1 | 80 | |||
AF213041.1 | - | 1 | 80 | |||
AF213046.1 | - | 1 | 80 | |||
AF213050.1 | - | 1 | 80 | |||
AF213052.1 | - | 1 | 80 | |||
AF213053.1 | - | 1 | 80 | |||
AK098653.1 | - | 2 | 194 | |||
AK293265.1 | - | 2 | 120 | |||
AY257474.1 | - | 1 | 80 | |||
AY257475.1 | - | 1 | 80 | |||
AY257476.1 | - | 1 | 80 | |||
AY257477.1 | - | 1 | 80 | |||
AY257478.1 | - | 1 | 80 | |||
BC103633.1 | - | 2 | 126 | |||
BT019451.1 | - | 1 | 80 | |||
CR407666.1 | - | 1 | 80 | |||
EF560750.1 | - | 2 | 120 | |||
L25876.1 | - | 2 | 141 | |||
L27711.1 | - | 2 | 131 | |||
U02681.1 | - | 2 | 111 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
