siRNA Details
Gene Symbol: C21orf91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100420.1 | NP_001093890.1 | 5 | 921 | ||
NM_001100421.1 | NP_001093891.1 | 4 | 858 | |||
NM_017447.3 | NP_059143.3 | 5 | 918 | |||
GenBank mRNA | AF239726.1 | - | 4 | 811 | ||
AK023825.1 | - | 5 | 886 | |||
AK225123.1 | - | 4 | 828 | |||
BC015468.2 | - | 4 | 834 | 3916462 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
