siRNA Details
Gene Symbol: C21orf91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017447.3 | NP_059143.3 | 4, 5 | 822 | ||
GenBank mRNA | BC015468.2 | - | 3, 4 | 738 | 3916462 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
