siRNA Details
Gene Symbol: RBM47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019027.3 | NP_061900.2 | 3, 4 | 1376 | ||
XM_017008310.1 | XP_016863799.1 | 4, 5 | 1513 | |||
GenBank mRNA | AK000280.1 | - | 3, 4 | 1354 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
