siRNA Details
Gene Symbol: CPVL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019029.2 | NP_061902.2 | 10 | 1143 | ||
NM_031311.3 | NP_112601.3 | 10 | 1055 | |||
XM_011515437.1 | XP_011513739.1 | 10 | 1222 | |||
GenBank mRNA | AF106704.2 | - | 10 | 975 | ||
AF217508.1 | - | 9 | 942 | |||
AF282617.1 | - | 10 | 1005 | |||
AK075433.1 | - | 10 | 1056 | |||
AK124472.1 | - | 14 | 1583 | |||
AK225545.1 | - | 10 | 1006 | |||
AY358549.1 | - | 10 | 1032 | |||
BC016838.2 | - | 10 | 1103 | 3888647 |
