siRNA Details
Gene Symbol: CCNJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019084.4 | NP_061957.2 | 4, 5 | 931 | ||
XM_017016355.1 | XP_016871844.1 | 3, 4 | 731 | |||
GenBank mRNA | BC043175.1 | - | 4, 5 | 941 | 5287365 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
