siRNA Details
Gene Symbol: CCNJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134375.1 | NP_001127847.1 | 5, 6 | 1098 | ||
NM_001134376.1 | NP_001127848.1 | 5, 6 | 1062 | |||
NM_019084.4 | NP_061957.2 | 5, 6 | 1065 | |||
XM_005269921.3 | XP_005269978.1 | 3, 4 | 538 | |||
XM_011539885.2 | XP_011538187.1 | 5, 6 | 1168 | |||
XM_017016354.1 | XP_016871843.1 | 4, 5 | 1368 | |||
XM_017016355.1 | XP_016871844.1 | 4, 5 | 865 | |||
XM_017016356.1 | XP_016871845.1 | 4, 5 | 862 | |||
GenBank mRNA | AK001757.1 | - | 2, 3 | 325 | ||
AK092360.1 | - | 5, 6 | 881 | |||
AK297264.1 | - | 5, 6 | 917 | |||
AK303018.1 | - | 2, 3 | 351 | |||
BC043175.1 | - | 5, 6 | 1075 | 5287365 | ||
BM789816.1 | - | 3, 4 | 648 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
