siRNA Details
Gene Symbol: BTN2A3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027795.1 | 3 | 632 | |||
GenBank mRNA | AK056871.1 | - | 3 | 612 | ||
AK292658.1 | - | 3 | 632 | |||
BC143636.1 | - | 2 | 388 | |||
DA706324.1 | - | 3 | 501 |
