siRNA Details
Gene Symbol: ZNF586 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077426.2 | NP_001070894.1 | 2 | 711 | ||
NM_001204814.1 | NP_001191743.1 | 4 | 888 | |||
NM_017652.3 | NP_060122.2 | 3 | 838 | |||
GenBank mRNA | AB074270.1 | - | 3 | 821 | ||
AK000077.1 | - | 3 | 819 | |||
AK095993.1 | - | 4 | 886 | |||
AK300419.1 | - | 2 | 668 | |||
AK302151.1 | - | 4 | 794 | |||
BC124558.1 | - | 2 | 649 | 40085152 |
