siRNA Details
Gene Symbol: BEST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017682.2 | NP_060152.2 | 6 | 730 | ||
XM_005259963.3 | XP_005260020.1 | 7 | 930 | |||
GenBank mRNA | AF440756.1 | - | 6 | 730 | ||
AK000139.1 | - | 6 | 665 | |||
AK225232.1 | - | 6 | 665 | |||
AY515705.1 | - | 6 | 730 |
