siRNA Details
Gene Symbol: FAM83E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017708.3 | NP_060178.2 | 4 | 877 | ||
XM_005259019.1 | XP_005259076.1 | 5 | 2201 | |||
GenBank mRNA | AK000207.1 | - | 4 | 1003 | ||
BC111970.1 | - | 4 | 877 | 8327438 | ||
BC111972.1 | - | 4 | 877 | 8327440 |
