siRNA Details
Gene Symbol: TMEM104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321264.1 | NP_001308193.1 | 6 | 533 | ||
NM_017728.3 | NP_060198.3 | 6 | 533 | |||
XM_006721963.1 | XP_006722026.1 | 6 | 507 | |||
XM_017024797.1 | XP_016880286.1 | 6 | 472 | |||
XM_017024798.1 | XP_016880287.1 | 6 | 507 | |||
XM_017024799.1 | XP_016880288.1 | 6 | 507 | |||
GenBank mRNA | AK000262.1 | - | 6 | 516 | ||
AK074029.1 | - | 6 | 493 | |||
AK291159.1 | - | 6 | 528 | |||
AK296621.1 | - | 6 | 556 | |||
BC037278.1 | - | 6 | 485 | 4825529 | ||
DN999695.1 | - | 6 | 539 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
